missing translation for 'onlineSavingsMsg'
Learn More

Whatman™ UniPrep™ Syringeless Filters - PVDF

Process samples in one-third the time of conventional syringe filters

Brand:  Whatman™ UN113UAQU

Code : A0

Additional Details : Weight : 0.43000kg

 View more versions of this product

Product Code. 11343565

  • £174.72 / Pack of 50


Estimated Shipment
09-03-2018
Add to basket

Specifications

Specifications

Syringeless Filter
PVDF
0.45μm
17mm
50μL
0.28cm sq.
Videos