missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ UniFilter™ Microplates, 96-Well, 2mL
Use for biomolecular purification by solid-phase extraction and organic synthesis in combinatorial chemistry library generation
Brand: Cytiva 7700-7206
Code : 69
Additional Details : Weight : 4.05000kg
Description
- Glass-filled polypropylene microplates
- Patented integral filter design prevents well-to-well cross contamination
- Standard footprint—compatible with most plate readers, autosamplers and robots
- Unique drip director design on the bottom of the plates controls filtrate placement
- Available in several media types
Specifications
Standard | |
GE’s Whatman UNIFILTER 2 mL, 96 well microplates are used for large sample or reagent volumes, as in biomolecule purification by SPE and organic synthesis in combinatorial chemistry library generation. | |
0.45 μm | |
96 | |
Biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation | |
25/Pk. |
Hydrophilic PVDF | |
8 x 12 | |
Glass Filled Polypropylene | |
Filter | |
Long drip director |