Learn More
Whatman™ UniFilter™ Microplates, 96-Well, 2mL
Use for biomolecular purification by solid-phase extraction and organic synthesis in combinatorial chemistry library generation
Brand:  Whatman™ 7700-7201
Code : 69
Additional Details : Weight : 3.98000kg
 View more versions of this product
Product Code. 11936886
  • £587.69 / Pack of 25
Estimated Shipment
05-06-2018
Add to basket
Description
Description
  • Glass-filled polypropylene microplates
  • Patented integral filter design prevents well-to-well cross contamination
  • Standard footprint—compatible with most plate readers, autosamplers and robots
  • Unique drip director design on the bottom of the plates controls filtrate placement
  • Available in several media types
Specifications
Specifications
Filtering Microplate
96 well
Glass Filled Polypropylene
96
Use for biomolecular purification by solid-phase extraction and organic synthesis in combinatorial chemistry library generation, sample preparation, genomics, filter-based assays and SPE
Long drip director
GF/C
2000μL
Videos
Documentation
Technical Documents