missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ UniFilter™ Microplates, 96-Well, 2mL
Use for biomolecular purification by solid-phase extraction and organic synthesis in combinatorial chemistry library generation
£690.00
Specifications
Format | Standard |
---|---|
Description | GE’s Whatman UNIFILTER 2 mL, 96 well microplates are used for large sample or reagent volumes, as in biomolecule purification by SPE and organic synthesis in combinatorial chemistry library generation. |
Array | 8 x 12 |
No. of Wells | 96 |
Well Shape | Filter |
Product Code | Brand | Membrane | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Membrane | Price | Quantity & Availability | |||||
11975114
|
Cytiva
7700-7206 |
Hydrophilic PVDF |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Glass-filled polypropylene microplates
- Patented integral filter design prevents well-to-well cross contamination
- Standard footprint—compatible with most plate readers, autosamplers and robots
- Unique drip director design on the bottom of the plates controls filtrate placement
- Available in several media types
Specifications
Standard | |
8 x 12 | |
Filter | |
Long drip director |
GE’s Whatman UNIFILTER 2 mL, 96 well microplates are used for large sample or reagent volumes, as in biomolecule purification by SPE and organic synthesis in combinatorial chemistry library generation. | |
96 | |
Biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation | |
25/Pk. |