missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ UniFilter™ 96-Well, 800μL Microplates

Suited for purifications, isolations, and separation of biomolecules, such as DNA

Brand:  Cytiva 7700-2805

Code : A0

Additional Details : Weight : 2.69200kg

 View more versions of this product

Product Code. 11977348

  • £665.00 / Pack of 25
Estimated Shipment: 10-05-2024
to see stock.
Add to basket

Description

Description

Suited for purifications, isolations, and separation of biomolecules, such as DNA

  • Clear polystyrene plates available with a variety of filter media
  • Integral filter design prevents well-to-well crosstalk
  • Unique drip director design on the bottom of the plates controls filtrate placement
    • Long drip directors eliminate well-to-well contamination during the filtration process
    • Short drip directors are effective for screening or filtering to waste
  • Standard footprint is compatible with most plate readers, autosamplers, and robots

Specifications

Specifications

Standard
UNIFILTER 96-well microplate, 800 μl, long drip director
0.45 μm
Polystyrene
Filter
Long drip director
Polypropylene 0.45μm
8 x 12
Clear
96
Typically used in purifications, isolations, and separations of biomolecules, particularly DNA
25/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers