Learn More
Cytiva Whatman™ UniFilter™ 384-Well Microplates
Microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples.
Brand: Cytiva 7700-2106
Additional Details : Weight : 2.24000kg
Description
Microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples.
Specifications
Standard | |
GE’s Whatman UNIFILTER 384 well microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples. | |
Clear | |
384 | |
Used for DNA template clean-up, cell capture, and for the removal of unwanted debris |
0.45 μm Hydrophilic PVDF | |
16 x 24 | |
Polystyrene | |
Filter | |
Long drip director |