Learn More
Cytiva Whatman™ Type WCN Cellulose Nitrate Membranes
Recommended for majority of routine applications, this membrane is manufactured under strictly controlled conditions. Whatman™ Type WCN Cellulose Nitrate Membranes narrow pore size distribution for improved surface capture and analysis.
Brand: Cytiva 7190-002
180.74 GBP valid until 2024-12-31
Use promo code "21931" to get your promotional price.
Code : A0
Additional Details : CAS Number : 9004-70-0 Weight : 0.06660kg
Description
Recommended for majority of routine applications, this membrane is manufactured under strictly controlled conditions. Whatman™ Type WCN Cellulose Nitrate Membranes narrow pore size distribution for improved surface capture and analysis.
- Among the strongest of their type with noticeably improved flexibility and tolerating abuse without sacrificing integrity
- Low level of extractables—generally below that of other similar membranes
- Advanced manufacturing ensures narrow pore size distribution and minimal batch-to-batch variation
- Autoclavable at 121°C (250°F), with low shrinkage
- Gridded circles available (Cat. No. 09-529-710 Series)
Specifications
Autoclavable | |
Cellulose Nitrate | |
Non-sterile | |
Hydrophilic | |
25 mm |
White | |
1.0 μm | |
Clarifying filtration (General) | |
100/Pk. |
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 1
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose