Learn More
Cytiva Whatman™ TurboBlotter Transfer System, Small Kits
Downward blotting device turns the conventional Northern/Southern transfer stack upside down for rapid, high-resolution transfer of DNA and RNA
Brand: Cytiva 10416328
Code : 69
Additional Details : Weight : 0.90000kg
Includes: Includes device plus five sets of transfer materials for gel size indicated in ordering table. Each set consists of one sheet of Nytran™ SPC 0.45μm Nylon membrane, one wick of 3MM Chr blotting paper and a blotter stack composed of eight sheets of 3MM Chr blotting paper and 20 sheets of GB004 blotting paper. Refills, available separately, contain five sets of transfer materials.
Description
- Takes advantage of gravity — no heavy weights are required on the top of the transfer stack, eliminating the messy set-up of standard upward capillary transfers
- Has a smaller footprint than most homemade devices and is stackable — up to five units can be stacked during transfers
- Allows for alkaline buffer transfers in one hour and neutral (SSC) transfers in three hours
- Very easy to set up and works without power or vacuum source
- Requires less buffer and blotting paper than conventional transfers
- Reusable
- Model is 12 × 16cm, for gels from 7 × 8cm to 11 × 14cm
Order Info
Refill packs containing additional membranes and papers required for further transfer are available separately (Cat. Nos. 09-301-177, -179, -181, -183, -185 and -189).
Specifications
DNA/RNA Transfer | |
Nytran™ SPC 0.45μm TurboBlotter™ Kit | |
3.9 in. | |
2.7 in. |
7 x 10 cm Gel TurboBlotter™ Transfer System | |
12 x 16cm Small transfer device, 5 sets of transfer materials (set consists of one sheet of Nytran™ SPC 0.45μm Nylon membrane, one wick of 3mm chromatography blotting Paper and a blotter stack composed of eight sheets o | |
10 cm | |
7 cm |