missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Syringe-Type Membrane Holder
Stainless steel membrane holder for use with a standard syringe
£167.00 - £386.00
Product Code | Brand | Material | Diameter (Metric) Inside | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Diameter (Metric) Inside | Price | Quantity & Availability | ||||
11380644
|
Cytiva
1980-002 |
Stainless Steel | 25 mm |
£386.00
Each |
Estimated Shipment: 17-04-2024
Log in to see stock. |
||||
11370644
|
Cytiva
1980-001 |
Stainless Steel | 13 mm |
£167.00
Each |
Estimated Shipment: 17-04-2024
Log in to see stock. |
||||
Description
Specifications
Stainless Steel |