missing translation for 'onlineSavingsMsg'
Learn More

Whatman™ SteriVent™ Vessel Venting Filter

For sterile venting of vessels and tanks

Brand:  Whatman™ 2107

Code : 69

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 11363425

  • £103.00 / Each


Estimated Shipment
09-03-2018
Add to basket

Specifications

Specifications

0.2μm
Yes
SB 0.375 to 0.5 in. (10 to 12mm)
Capsule 150 (Vented)
1/pk.
2000cm2
Videos