Learn More
Cytiva Whatman™ Sterile Mixed Cellulose Ester Membranes, ME 25
Cytiva Whatman Sterile Mixed Cellulose Ester Membranes, ME 25, Black, Diameter Outside: 47mm, 1.85 in., Grid Color: White, Autoclavable: Yes, Grade: ME 25, Includes: With interleaving paper and without pad, Membrane Type: WME, Pore Size: 0.45um, Protein Binding Capacity: Medium
Brand: Cytiva 10406803
Code : A0
Additional Details : Weight : 0.03800kg
Description
Cytiva Whatman Sterile Mixed Cellulose Ester Membranes, ME 25, Black, Diameter Outside: 47mm, 1.85 in., Grid Color: White, Autoclavable: Yes, Grade: ME 25, Includes: With interleaving paper and without pad, Membrane Type: WME, Pore Size: 0.45um, Protein Binding Capacity: Medium
Specifications
Autoclavable | |
White | |
ME 25 | |
Sterile | |
Black | |
Medium | |
130°C | |
47 mm |
4.3 to 5.0 mg/cm2 | |
Mixed Cellulose Ester | |
0.45 μm | |
membrane butler | |
Without interleaving paper and pad | |
3.1mm | |
Membrane |
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 1
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose