missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Reeve Angel RA TCLP Glass Fiber Filters
Assists with determining the leaching potential in a landfill for hazardous organic and inorganic contaminants into groundwater threatening drinking water
Brand: Cytiva 5925-090
Code : A0
Additional Details : Weight : 0.01100kg
Description
- Material: borosilicate glass circles
- Diameter: 90mm (3.5 in.)
- Pore size: 0.6-0.8 μm
Specifications
Borosilicate Glass Fiber | |
0.6 to 0.8 μm | |
RA TCLP | |
90 mm |