missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Reeve Angel 802 Prepleated Qualitative Circles
Coarse retention
Brand: Cytiva 5802-240
Code : 69
Additional Details : Weight : 0.01100kg
Description
- Folded for fast filtration of coarse particles and gelatinous precipitates
- Qualitative Grade; nominal retention 30-35μm; 0.26mm thick Pleating and creped surface increase effective area, improving both filtering speed and extending filter life
- Wet strengthened†
Specifications
Disc | |
30 to 35 μm | |
Coarse | |
24 cm |
15 μm | |
802 | |
0.26 mm |