missing translation for 'onlineSavingsMsg'
Learn More

Whatman™ Reeve Angel 802 Prepleated Qualitative Circles

Coarse retention

Brand:  Whatman™ 5802-185

Code : 69

Additional Details : Weight : 0.22000kg

 View more versions of this product

Product Code. 11781598

  • £23.22 / Pack of 100


Estimated Shipment
10-05-2018
Add to basket

Description

Description

  • Folded for fast filtration of coarse particles and gelatinous precipitates
  • Qualitative Grade; nominal retention 30-35μm; 0.26mm thick Pleating and creped surface increase effective area, improving both filtering speed and extending filter life
  • Wet strengthened†

Specifications

Specifications

18.5cm
30 to 35μm
15μm
0.26mm
802
Coarse
Circle
Videos