Learn More
Cytiva Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters
Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.
Brand: Cytiva 6788-2504
432.61 GBP valid until 2024-05-06
Use promo code "21711" to get your promotional price.
Code : 75
Additional Details : Weight : 0.55440kg
Description
Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.
- Diameter: 25mm
- Sample volume up to 100mL
- Low hold-up volumes for maximum sample recovery
- Optional glass microfiber
- Autoclave at 121ºC
- HPLC aqueous sample preparation
- Biological sample preparation
- Buffer solutions
- Salt solutions
- Tissue culture media
- Irrigation solutions
- Sterile isolation
Specifications
Puradisc 25 Depth Polypropylene Syringe Filter, 0.45 μm (200 pcs) | |
0.45 μm | |
4.2 cm2 | |
Female luer lock | |
Male luer | |
Syringe Filter | |
22.9 x 28.4 mm |
Depth Polypropylene (DpPP) | |
Non-sterile | |
Pigment Free Polypropylene (PP) | |
200 | |
5.2 bar (75 psi) | |
Up to 100 mL | |
25 mm |