missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters

Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.

Brand:  Cytiva 6786-2504

Code : 75

Additional Details : Weight : 0.16650kg

 View more versions of this product

Product Code. 11314694

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.

  • Diameter: 25mm
  • Sample volume up to 100mL
  • Low hold-up volumes for maximum sample recovery
  • Optional glass microfiber
  • Autoclave at 121ºC
Applications:
  • HPLC aqueous sample preparation
  • Biological sample preparation
  • Buffer solutions
  • Salt solutions
  • Tissue culture media
  • Irrigation solutions
  • Sterile isolation
Specifications

Specifications

Puradisc 25 Depth Polypropylene Syringe Filter, 0.45 μm (50 pcs)
0.45 μm
4.2 cm2
Female luer lock
Male luer
Syringe Filter
22.9 x 28.4 mm
Polypropylene
Non-sterile
Pigment Free Polypropylene (PP)
50
5.2 bar (75 psi)
Up to 100 mL
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers