missing translation for 'onlineSavingsMsg'
Learn More

Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.45μm BA85 Protran Circles

100% pure nitrocellulose to ensure the highest binding capacity possible

Brand:  Whatman™ 10401164

Code : 69

Additional Details : Weight : 0.08000kg

 View more versions of this product

Product Code. 11335294

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Universal membrane suitable for Western, Southern, and Northern blotting
  • Hydrophilic, so no prewetting with methanol is required
  • High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
  • Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
  • Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
  • Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa

Specifications

Specifications

50/Pk.
Nitrocellulose
87mm
0.45μm
Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection
High
Circle
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Documentation