missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.45μm BA85 Protran Circles
100% pure nitrocellulose to ensure the highest binding capacity possible
Brand: Cytiva 10401124
Code : 69
Additional Details : Weight : 0.11000kg
Description
- Universal membrane suitable for Western, Southern, and Northern blotting
- Hydrophilic, so no prewetting with methanol is required
- High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
- Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
- Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
- Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa
Specifications
Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection | |
132 mm | |
Disc | |
0.45 μm |
High | |
25/Pk. | |
Nitrocellulose |