missing translation for 'onlineSavingsMsg'
Learn More

Whatman™ Polydisc Inline Filters

Versatile, cost-effective filters designed for larger volume sample filtration

Brand:  Whatman™ 6721-5010

Code : 75

Additional Details : Weight : 0.19000kg

 View more versions of this product

Product Code. 11334634

  • £132.00 / Pack of 10


Estimated Shipment
15-06-2018
Add to basket

Description

Description

  • Filter up to 1L with one device
  • Can be used in conjunction with a syringe or connected inline (6-10mm I.D. hose) via stepped hose barbs
  • High-purity polypropylene housing maintains sample purity
  • 50mm filter devices
  • Autoclavable
  • Lightweight (11.5g)
Polydisc AS (Aqueous Solution)
  • For tissue culture media, reagent preparation, particle counting solutions, pharmaceutical preparations High-throughput polyethersulfone membrane (low protein binding and no surfactants)
  • Glass microfiber prefilter extends membrane life and effectively filters heavily contaminated samples
  • Sterility cap on outlet (optional)
  • No EtO residuals Barbed hose connections fit multiple tubing sizes Integrity testable (bubble point method) Sealed in its own “medical grade” clear blister pack and secured in a protective shelf pack
Polydisc TF
  • Use in vents, inline, exhausts for growth environments, clean or sterile gases, solvents, reagents, drying gases, photoresists Hydrophobic PTFE membrane ideal for chemically aggressive solutions, reagents and organic solvents
  • Chemical-resistant housing Integrity testable (bubble point or water breakthrough pressure in situ methods) Biosafe

Specifications

Specifications

1μm
Yes
Larger volume sample filtration
Polypropylene
Pk. of 10
Polydisc TF
Videos
Documentation