missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Polydisc Inline Filters

Versatile, cost-effective filters designed for larger volume sample filtration

Brand:  Cytiva 6720-5045

Code : 75

Additional Details : Weight : 0.19000kg

 View more versions of this product

Product Code. 11324634

  • £127.00 / Pack of 10
Estimated Shipment: 18-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Versatile, cost-effective filters designed for larger volume sample filtration

  • Filter up to 1L with one device
  • Can be used in conjunction with a syringe or connected inline (6-10mm I.D. hose) via stepped hose barbs
  • High-purity polypropylene housing maintains sample purity
  • 50mm filter devices
  • Autoclavable
  • Lightweight (11.5g)
Polydisc AS (Aqueous Solution)
  • For tissue culture media, reagent preparation, particle counting solutions, pharmaceutical preparations High-throughput polyethersulfone membrane (low protein binding and no surfactants)
  • Glass microfiber prefilter extends membrane life and effectively filters heavily contaminated samples
  • Sterility cap on outlet (optional)
  • No EtO residuals Barbed hose connections fit multiple tubing sizes Integrity testable (bubble point method) Sealed in its own “medical grade” clear blister pack and secured in a protective shelf pack
Polydisc TF
  • Use in vents, inline, exhausts for growth environments, clean or sterile gases, solvents, reagents, drying gases, photoresists Hydrophobic PTFE membrane ideal for chemically aggressive solutions, reagents and organic solvents
  • Chemical-resistant housing Integrity testable (bubble point or water breakthrough pressure in situ methods) Biosafe

Specifications

Specifications

Autoclavable
PTFE
16 cm2
Polypropylene (PP)
Pk. of 10
50 mm
Polydisc TF Filter, polypropylene, 0.45 μm (10 pcs)
0.45 μm
Larger volume sample filtration
Tubing Nozzle 6 to 10 mm i.d. hose
Polydisc TF
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers