missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Polydisc In-line Filter
For serum samples
Brand: Cytiva 6724-5000
152.98 GBP valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Code : 75
Additional Details : Weight : 0.19000kg
Description
- Glass microfiber and polyethersulfone membranes
- Contains stack of filter media
- Radiation sterilized
Specifications
PES | |
500.0mL/min./cm sq. | |
10/Pk. |
1.0 μm | |
For serum samples; In virology, microbiology, immunoassay |