missing translation for 'onlineSavingsMsg'
Learn More

Whatman™ 47mm Nuclepore™ Polycarbonate Track-Etched Membranes

Manufactured from high-quality polycarbonate film and track-etched for high pore size precision. Whatman™ 47mm Nuclepore™ Polycarbonate Hydrophilic Membranes combine high flow rates and chemical and thermal resistance with sharply defined pores, making them suitable for microscopy, cytology, parasitology, and certain air and oceanography analyses.

Brand:  Whatman™ 111113

Code : 50

Additional Details : Weight : 0.06000kg

 View more versions of this product

Product Code. 11302905

  • £91.65 / Pack of 100


Estimated Shipment
18-07-2018
Add to basket

Description

Description

Effective chemotaxis requires a delicate balance between cell adhesion and migration. Nucleopore Membranes combine PVP-free polypropylene with sharp, uniform cylindrical pores to achieve both.
  • Offers a highly defined pore size cutoff in the sub-micron range, enabling particle deformity measurement and capture of extremely small particles
  • Track-etched for highly defined pore size and high pore density
  • Treated with PVP to render it hydrophilic
  • Low extractables, minimizing sample contamination risk
  • High chemical resistance and good thermal stability for a wide range of sample types
  • Low, consistent ash and low tare weights
  • Smooth flat surface for high particle visibility
Specifications

Specifications

5μm
47mm
Hydrophilic
Nuclepore Polycarbonate
Epifluorescence microscopy, environmental analysis, cell biology
High
Videos
Documentation