Learn More
Cytiva Whatman™ Grade 8 Ruled Special-Purpose Filter Paper
Hardened grade for routine analysis of extraneous matter in food, beverage and pharmaceutical laboratories
Brand: Cytiva 10347008
Code : A0
Additional Details : Weight : 0.04000kg
Description
™Resistant to aqueous and non-aqueous solutions
Recovers and measures contaminants in food products™
Specifications
Cotton Linters | |
5mm green rules are resistant to aqueous, non-aqueous solution | |
2.75 in. |
Qualitative analytical techniques to determine and identify materials | |
Ruled Special Purpose Filter Paper | |
7 cm |