missing translation for 'onlineSavingsMsg'
Learn More

Whatman™ Grade 598 1/2 Qualitative Folded Paper Filter - Circle

Decreased overall filtration time and increased total loading capacity

Brand:  Whatman™ 10312256

Code : A0

Additional Details : Weight : 1.58000kg

 View more versions of this product

Product Code. 11383055

  • £197.60 / Pack of 50


Estimated Shipment
07-02-2018
Add to basket

Specifications

Specifications

50/Pk.
598.5, Folded (Prepleated)
Medium Fast, Thick
320μm
500mm
8 - 10μm
Folded
Videos