missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Grade 287 1/2 Folded Filter Paper
Folded filter paper saves time, increases loading capacity
£44.30 - £65.30
Specifications
Filtration Speed | Approx. 330 s |
---|---|
For Use With (Application) | For clarifying milk serum, starch solutions, soil suspensions or sugar-containing solutions prior to polarimetry or refractometry |
Grade | Grade 287 ½ |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11338894
|
Cytiva
10310247 |
185 mm |
£65.30
Pack of 50 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
11328894
|
Cytiva
10310245 |
150 mm |
£54.80
Pack of 50 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
11318894
|
Cytiva
10310244 |
125 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Kieselguhr paper with medium to slow flow
- For separation of very fine semi-colloidal turbidity
- For clarifying milk serum, starch or sugar-coated solutions, soil suspensions
Specifications
Approx. 330 s | |
For clarifying milk serum, starch solutions, soil suspensions or sugar-containing solutions prior to polarimetry or refractometry | |
Grade 287 ½ |