missing translation for 'onlineSavingsMsg'
Learn More

Whatman™ Grade 0905 Qualitative Creped Filter Paper Sheets

Designed for applications ranging from the filtration of beverages to the purification of electroplating baths

Brand:  Whatman™ 10334985

Code : A0

Additional Details : Weight : 8.00000kg

 View more versions of this product

Product Code. 11348804

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Very high filtration speed
  • Creped for coarse particles

Specifications

Specifications

0905
Super-refined Cellulose
12 to 25μm
Sheet
450mm
450mm
High
Videos