missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Grade 0858 Filter Papers for Technical Use
Grained, with medium fast flow rate and medium retention. A universal filter paper, used for the filtration of extracts, oils, beer, syrups etc.; also applied in filter presses or for the aspiration of liquids.
Brand: Cytiva 10334365
Code : 69
Additional Details : Weight : 2.24000kg
Description
These filter papers are made from super-refined cellulose and have been specifically designed to have particular properties for each technical application. The applications range from the filtration of beverages to the purification of electroplating baths.Specifications
Super-refined Cellulose | |
0858 | |
4 x 23 in. (11 x 58cm) | |
11 cm | |
58 cm |
Medium | |
170 μm | |
4 in. | |
23 in. |