missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Grade 0858 Filter Papers for Technical Use

Grained, with medium fast flow rate and medium retention. A universal filter paper, used for the filtration of extracts, oils, beer, syrups etc.; also applied in filter presses or for the aspiration of liquids.

£161.00 - £393.00

Specifications

Flow Rate Medium
Thickness (Metric) 170 μm
View More Specs

Products 3
Product Code Brand Length (English) Length (Metric) Width (English) Width (Metric) Price Quantity & Availability  
Product Code Brand Length (English) Length (Metric) Width (English) Width (Metric) Price Quantity & Availability  
11368794
View Documents Promotion Details
Cytiva
10334383
15.35 in. 39 cm 15.35 in. 39 cm
£316.00
Pack of 500
View alternative products
Save Up to 
»
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
 
11358794
View Documents Promotion Details
Cytiva
10334365
4 in. 11 cm 23 in. 58 cm
£161.00
Pack of 500
View alternative products
Save Up to 
»
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
 
10653772
View Documents Promotion Details
Cytiva
10334385
17.72 in. 45 cm 17.72 in. 45 cm
£393.00
Pack of 500
View alternative products
Save Up to 
»
Estimated Shipment: 17-06-2024
to see stock.
Add to basket
 
Description

Description

These filter papers are made from super-refined cellulose and have been specifically designed to have particular properties for each technical application. The applications range from the filtration of beverages to the purification of electroplating baths.
Specifications

Specifications

Medium
170 μm
Videos
SDS
Documents

Documents

Product Certifications