Learn More
QIAGEN QIAcard FTA Gene
For collection, stabilization, processing, transport and archiving of biological samples. 3 sample areas per card.
Brand: QIAGEN WB120208
Code : 69
Additional Details : Weight : 0.51000kg
Description
For collection, stabilization, processing, transport and archiving of biological samples. 3 sample areas per card.
- Three sample areas in a chipboard frame suitable for several automated dispensing/pipetting or punching systems.
- Storage of up to 75 μL of blood (whole blood, EDTA, sodium citrate, ACD, or heparin anticoagulated blood) to each printed circle of each FTA Gene Card (total 225 μL per card).
- Apply only one blood sample to each card.
Specifications
FTA Gene Card, 3 sample areas per card, 75 μl max volume/sample area, 225 μl max total volume/card | |
3 | |
100 Pk. |
Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology | |
FTA Gene Card | |
75 μL |
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4