Learn More
Whatman™ Elutrap™ Electroelution Membranes
Designed to isolate nucleic acids and proteins from agarose or polyacrylamide gel slices
Brand: Whatman™ 10404092
Code : A0
Additional Details : Weight : 0.10000kg
Description
- For most horizontal gel electrophoresis chambers
- Efficient current flow
- Handles up to four samples simultaneously
- Purifies nucleic acids 14bp to 150Kb; proteins larger than 3 to 5kD
- No salt cushions or special buffers required
- Adjustable trap allows optimization of final sample volume
Specifications
Elutrap™ electroelution system | |
BT2 membrane | |
Eluting nucleic acids (14 bp to 150Kb) and proteins (larger than 3 to 5kD) from gel slices |