Learn More
Cytiva Whatman™ Cellulose Nitrate Membrane Filters
Manufactured under strictly controlled conditions. Whatman™ Cellulose Nitrate Membrane Filters have a noticeably improved flexibility and are made to tolerate abuse during handling, loading, and autoclaving without sacrificing integrity.
Brand: Cytiva 10401306
Code : A0
Additional Details : Weight : 0.01100kg
Description
- Narrow pore size distribution for improved surface capture and analysis.
- Low levels of extractables to ensure sample integrity.
- High strength and flexibility
- Increased temperature stability
- Reduced shrinkage
Specifications
Disc | |
0.2 μm | |
105 to 140 μm | |
25 mm |
Cellulose Nitrate | |
100 Pack | |
1.9 in. |
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.