missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Binder-Free Glass Microfiber Filters, Grade 934-AH Circles
Designed for analysis of non-filterable residue and materials that require a high loading capacity glass microfiber filter. Whatman™ Binder-Free Glass Microfiber Filters, Grade 934-AH Circles are also used in cell harvesting, liquid scintillation, and monitoring pollution.
£25.70 - £785.00
Specifications
Basis Weight | 64 g/m2 |
---|---|
Description | Whatman Grade 934-AH borosilicate glass microfiber filters from GE are smooth water quality filters with high retention efficiency. Grade 934-AH filters are often used in total suspended solids analysis. |
Format | Disc |
Material | Borosilicate Glass |
Particle Retention | 1.5 μm |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11334894
|
Cytiva
1827-021 |
21 mm |
£25.70
Pack of 100 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
10471602
|
Cytiva
1827-024 |
24 mm |
£25.80
Pack of 100 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
|||||
11354894
|
Cytiva
1827-025 |
25 mm |
£25.80
Pack of 100 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
|||||
11374894
|
Cytiva
1827-028 |
28 mm |
£26.55
Pack of 100 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
11384894
|
Cytiva
1827-030 |
30 mm |
£34.75
Pack of 100 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
11304904
|
Cytiva
1827-032 |
32 mm |
£30.90
Pack of 100 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
11314904
|
Cytiva
1827-035 |
35 mm |
£35.05
Pack of 100 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
11324904
|
Cytiva
1827-037 |
37 mm |
£40.20
Pack of 100 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
|||||
11334904
|
Cytiva
1827-042 |
42.5 mm |
£37.05
Pack of 100 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
11370614
|
Cytiva
1827-047 |
47 mm |
£67.65
Pack of 100 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
11380614
|
Cytiva
1827-055 |
55 mm |
£77.65
Pack of 100 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
11390614
|
Cytiva
1827-070 |
70 mm |
£135.00
Pack of 100 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
11354904
|
Cytiva
1827-082 |
82 mm |
£168.00
Pack of 100 |
Estimated Shipment: 01-05-2024
Log in to see stock. |
|||||
11364904
|
Cytiva
1827-085 |
85 mm |
£115.00
Pack of 100 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
10022171
|
Cytiva
1827-090 |
90 mm |
£146.00
Pack of 100 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
|||||
11304894
|
Cytiva
1827-105 |
105 mm |
£139.00
Pack of 100 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
11310624
|
Cytiva
1827-110 |
110 mm |
£196.00
Pack of 100 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
11320624
|
Cytiva
1827-125 |
125 mm |
£236.00
Pack of 100 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
11314894
|
Cytiva
1827-150 |
150 mm |
£240.00
Pack of 100 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
11324894
|
Cytiva
1827-185 |
185 mm |
£279.00
Pack of 100 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
11344894
|
Cytiva
1827-240 |
240 mm |
£469.00
Pack of 100 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
11394894
|
Cytiva
1827-320 |
320 mm |
£785.00
Pack of 100 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
Description
Designed for analysis of non-filterable residue and materials that require a high loading capacity glass microfiber filter. Whatman™ Binder-Free Glass Microfiber Filters, Grade 934-AH Circles are also used in cell harvesting, liquid scintillation, and monitoring pollution.
Grade 934-AH filters are often used in total suspended solids analysis.- Smooth surface for efficient precipitate recovery
- Nominal particle retention of 1.5μm
- Withstands temperatures up to 550°C
- Fast filtration speed
Specifications
64 g/m2 | |
Disc | |
1.5 μm | |
3.7 sec./100mL/in.2 | |
100 Pack | |
Smooth | |
0.017 in. | |
Filter for Total Suspended Solids Analysis |
Whatman Grade 934-AH borosilicate glass microfiber filters from GE are smooth water quality filters with high retention efficiency. Grade 934-AH filters are often used in total suspended solids analysis. | |
Borosilicate Glass | |
47 sec./100mL | |
Grade 934-AH | |
Circle | |
932°F | |
435 μm | |
341 mL/min |