Learn More
Cytiva Whatman™ Autovial™ Syringeless Filters: 12mL Capacity, Nonsterile
Faster sample handling, up to 12mL in volume
Brand: Cytiva AV125UPP
Code : A0
Additional Details : Weight : 0.62000kg
Description
Replace syringe-coupled devices with single filter units for one-time use. Often used for removing particulates from potentially hazardous samples. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial.
- Preassembled units consist of a filter barrel and a plunger
- Built-in air purge minimizes holdup
- Integrated construction eliminates filter pop-off
- Support stand protects slip-Luer tip, recessed to reduce chance of contamination
- Handle samples up to 12mL in volume
- All have an integral 25mm diameter filter in barrel (graduated in 1mL intervals) with polyethylene plunger
- Filtration area: 3.0cm2
- Glass microfiber prefilter ensures higher flow with less clogging
- Units with NYL, PTFE, and PVDF membranes are available sterile
Cellulose Acetate (CA)
- Low nonspecific protein binding, high loading capacity; for biologicals
- High loading capacity, binder-free, fast flowrates; for clarifying aqueous or organic solutions, pH 3 to 8
- Inherently hydrophilic; for clarifying aqueous and organic samples, pH 3 to 10
- For aqueous and organic solutions; ideal for aggressive samples and high debris
- Hydrophobic; for samples >50% organic solvents
- Low protein binding; for aqueous samples
Specifications
Polypropylene | |
Non-sterile | |
Syringeless Filter | |
25 mm |
0.45 μm | |
50/Pk. | |
≤12 mL |