missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare, Whatman Anotop 25mm HPLC Syringe Filters, Pore size 0.2μm

Whatman Anotop HPLC syringe filter products from GE Healthcare’s Life Sciences business are specifically designed for effective UHPLC and HPLC sample preparation.

Brand:  GE Healthcare, Whatman 2002-5100

Code : 75

Additional Details : Weight : 0.47000kg

 View more versions of this product

Product Code. 11320654

  • £824.00 / Pack of 100


Estimated Shipment
07-06-2018
Add to basket

Description

Description

  • Syringe filter removes particulates that could cause damage, supporting longer column life
  • Low levels of UV-absorbing extractable substances for consistent HPLC sample preparation
  • Anopore inorganic aluminum membrane filters a broad range of solvents
  • High pore-size precision due to capillary pore structure
Specifications

Specifications

Syringe Filter
Proprietary Alumina Based Anapore Membrane
100/Pk.
No
2.5cm
100mL
0.2μm
Videos