missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Anotop™ 25 Non-Sterile Syringe Filters
Remove virtually 100% of particulates larger than pore size
£625.00 - £2370.00
Specifications
Disposable | Single-use |
---|---|
Material (Membrane) | Anopore Inorganic Membrane (alumina matrix) |
Sterility | Non-sterile |
Filtration Area | 4.78 cm2 |
For Use With (Application) | HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water |
Product Code | Brand | Pore Size | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Quantity | Price | Quantity & Availability | ||||
11364714
|
Cytiva
6809-2024 |
0.2 μm | 200/Pk. |
£2370.00
Pack of 200 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
||||
11344714
|
Cytiva
6809-2012 |
0.1 μm | 50/Pk. |
£625.00
Pack of 50 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
||||
11354714
|
Cytiva
6809-2022 |
0.2 μm | 50/Pk. |
£829.00
Pack of 50 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
11334714
|
Cytiva
6809-2002 |
0.02 μm | 50/Pk. |
£845.00
Pack of 50 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
||||
Description
Compatible with aqueous suspensions and a wide range of organic solvents. Handle volumes up to 5mL (10mm diameter filter) or 100mL (25mm diameter filter).
- Nondeformable, honeycombed Anopore™ membrane
- Inherently hydrophilic membrane is free of adhesives and surfactants
- Low protein binding, low fluid retention for maximum sample recovery
- Unidirectional flow
Order Info
Hexagonal, thermal-welded polypropylene housing is free of adhesives and binders.
Specifications
Single-use | |
Non-sterile | |
HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water | |
Polypropylene | |
Male Luer Slip | |
45°C | |
25 mm |
Anopore Inorganic Membrane (alumina matrix) | |
4.78 cm2 | |
<150 μL | |
Female Luer Lok | |
75psi | |
Syringe Filter |