Learn More
Cytiva Whatman™ Anotop™ 25 Non-Sterile Syringe Filters
Remove virtually 100% of particulates larger than pore size
Brand: Cytiva 6809-2002
684.37 GBP valid until 2024-05-06
Use promo code "21711" to get your promotional price.
Code : 75
Additional Details : Weight : 0.28980kg
Description
Compatible with aqueous suspensions and a wide range of organic solvents. Handle volumes up to 5mL (10mm diameter filter) or 100mL (25mm diameter filter).
- Nondeformable, honeycombed Anopore™ membrane
- Inherently hydrophilic membrane is free of adhesives and surfactants
- Low protein binding, low fluid retention for maximum sample recovery
- Unidirectional flow
Specifications
Single-use | |
0.02 μm | |
4.78 cm2 | |
<150 μL | |
Female Luer Lok | |
75psi | |
45°C | |
25 mm |
Anopore Inorganic Membrane (alumina matrix) | |
Non-sterile | |
HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water | |
Polypropylene | |
Male Luer Slip | |
50/Pk. | |
Syringe Filter |