Learn More
Cytiva Whatman™ Anotop™ 10mm Syringe Filters
Removes virtually 100% of particulates larger than pore size. Whatman™ Anatop™ 10mm Syringe Filters include a highly defined capillary pore structure and narrow pore-size distribution, enabling a high level of consistency in particle retention.
Brand: Cytiva 6809-1022
397.84 GBP valid until 2024-05-06
Use promo code "21711" to get your promotional price.
Code : A0
Additional Details : Weight : 0.05000kg
Description
Removes virtually 100% of particulates larger than pore size. Whatman™ Anatop™ 10mm Syringe Filters include a highly defined capillary pore structure and narrow pore-size distribution, enabling a high level of consistency in particle retention.
Compatible with aqueous suspensions and a wide range of organic solvents. Handle volumes up to 5mL (10mm diameter filter) or 100mL (25mm diameter filter).
- Nondeformable, honeycombed Anopore™ membrane
- Inherently hydrophilic membrane is free of adhesives and surfactants
- Low protein binding, low fluid retention for maximum sample recovery
- Unidirectional flow
Specifications
Single-use | |
0.2 μm | |
0.78 cm2 | |
<20 μL | |
Female Luer Lock | |
6.9bar (100psi) | |
45°C | |
Up to 10 mL |
Anopore Inorganic Membrane (alumina matrix) | |
Non-sterile | |
HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water | |
Polypropylene | |
Male Luer Slip | |
50 Pack | |
Syringe Filter | |
10 mm |