missing translation for 'onlineSavingsMsg'
Learn More

Whatman™ Anodisc™ Filter Membranes

Constructed with precision pore structure and narrow pore size distribution. Whatman™ Anodisc™ Filter Membranes ensure a high level of particle removal efficiency. Captures microorganisms and particulates on the surface of the membrane for subsequent analysis.

Brand:  Whatman™ 6809-6012

Code : A0

Additional Details : Weight : 2.15460kg

 View more versions of this product

Product Code. 11335044

  • £304.00 / Pack of 50


Estimated Shipment
12-06-2018
Add to basket

Description

Description

Anopore inorganic membrane circles available with or without support ring contain no lateral crossovers between individual pores.
  • Wide solvent compatibility reduces the need to stock a variety of membranes in the laboratory.
  • No additives used in the manufacturing process ensures minimal extractables and no sample contamination.
  • Extremely low protein binding minimizes sample loss.
  • Virtually transparent when wet making it highly suitable for microscopy studies.
Specifications

Specifications

0.1μm
25mm
No
Hydrophilic
Aluminum oxide
50Pack
Polypropylene support ring
Anodisc 25
Videos