Learn More
Cytiva Whatman™ 4 and 13mm Puradisc Polyethersulfone Syringe Filters
For minimal adsorptive protein loss
Brand: Cytiva 6781-1304
831.25 GBP valid until 2024-05-06
Use promo code "21711" to get your promotional price.
Code : 75
Additional Details : Weight : 0.47000kg
Description
Suitable for filtering serum, plasma and tissue culture solutions—as well as other protein-containing solutions where minimal adsorptive protein loss is required.
- Polypropylene housing
- Autoclavable
- Female Luer lock on inlet side; male slip Luer on outlet side
4mm dia. Filters
- For samples up to 2mL
- Filter area: 0.2cm2 Hold-up volume (with air purge) <7.5μL
- For samples up to 10mL
- Filter area: 1.3cm2
- Hold-up volume (after air purge) <25μL
Specifications
Autoclavable | |
Polysulfone | |
Non-sterile | |
≤25 μL | |
Male Luer Slip | |
Polypropylene | |
≤10 mL |
Passes USP Class VI requirements for biosafety. | |
0.45 μm | |
For filtering serum, plasma and tissue culture solutions – as well as other protein-containing solutions where minimal adsorptive protein loss is required. | |
Female Luer Lok | |
500/Pk. | |
Syringe Filter | |
13 mm |