Learn More
Cytiva Whatman™ 0.6 μm Pore Size (ME 26) Mixed Cellulose Ester Membranes, Plain
Whatman™ (ME 26) Mixed Cellulose Ester Membranes are particularly effective in applications requiring higher flow rates and larger volume filtration, including clarification or sterilization of aqueous solutions, particulate analysis and removal, air monitoring, and microbial analysis.
Brand: Cytiva 10401506
Code : 69
Additional Details : Weight : 0.00300kg
Description
- Filter Type: Plain
- Material: Mixed Cellulose Ester
- Material (Membrane): ME 26
- Pore Size: 0.6 μm
- Autoclavable: Yes
- Sterility: Non-sterile
- Flow Rate: 4.8 sec./100 mL/12.5 cm² (at Δp = 0.9 bar)
- Gas Flow Rate: 21 sec./100 mL (at Δp = 3 mbar)
- Bubble Point: 3.7 bar
- For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Specifications
Autoclavable | |
Single-use | |
MCE | |
0.6 μm | |
4.8 sec./100 mL/12.5cm² (at Δp = 0.9 bar) | |
21 sec./100 mL (at Δp = 3 mbar) | |
0.98 in. |
3.7 bar | |
Plain | |
ME 26 | |
Non-sterile | |
Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays | |
Plain | |
25 mm |
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 1
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose