Learn More
Thermo Scientific™ Wellwash™ Microplate Washer (EU IVD/CE-marked)
Wash 96-well plates securely with this easy-to-use, reliable microplate-strip washer for routine ELISA applications.
Brand: Thermo Scientific™ 5165060
Code : NEW
Additional Details : Weight : 8.00000kg
Alert:
Fulfills IVD Directive 98/79/EC requirements, valid only in Europe. Not for in vitro diagnostics use in North America.
Description
Wash 96-well plates securely with the Thermo Scientific™ Wellwash™ Microplate Washer, an easy-to-use, reliable microplate-strip washer for routine ELISA applications. The Wellwash features a graphical user interface, local language versions and USB port for optimal ease-of-use and convenience.
Simplify your ELISA workflow by using our eGuide to see our complete portfolio for all your ELISA needs.
Features:
- Large color clear display and keypad for easy, convenient use
- User interface is logic and intuitive for quick protocols development
- Easy-to-use software requires minimal training for fast startup
- Unpressurized bottles are safe and secure to use
- Liquid level sensors are present in both wash and waste bottle for security
- Automatic prime feature assure safe performance
- Aerosol cover prevents aerosolization of infectous diseases
- Plate sensor recognizes if plate is present
- Wellwash is designed to efficiently remove the excess liquid from the wells
- Low residual volumes provide optimal washing performance and reliable assay results
Includes:
1x2L wash bottle, 1x2L waste bottle, 1x8L wash head, aerosol cover
Recommended for:
ELISA-based assays
Warranty: 1 year
Specifications
< 5% @ 300μL | |
USB | |
Wash Bottle (2 L), Waste Bottle (2 L) | |
50 to 400 μL | |
96-Well Plates | |
8 kg | |
LCD | |
50/60 Hz | |
No | |
15.2 in. | |
13.6 in. | |
9.4 in. | |
No | |
100/240 V 50/60 Hz | |
5 to 100 mL |
CE Marked | |
1 | |
< 3% (CV) @ 300μL (96-well plate) | |
1 x 8 | |
<2 μL | |
50 to 1000 μL | |
ELISA-Based Assays | |
Wellwash 1x8 IVD model (Not available in USA) | |
1 x 8 wash head, 1 x 2L wash bottle, 1 x 2L waste bottle, aerosol cover | |
385 mm | |
345 cm | |
240 mm | |
17.6 lb. | |
100/240 V |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. Fulfills IVD Directive 98/79/EC requirements, valid only in Europe. Not for in vitro diagnostics use in North America.