Learn More
Thermo Scientific™ Wellcolex™ Color Salmonella Rapid Latex Agglutination Test Kit
Save up to two days over traditional screening methods with this easy-to-use, cost effective latex agglutination test for rapid, accurate identification of Salmonella.
£550.00 - £1440.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Description | Wellcolex Colour Salmonella Kit |
CE Marker | Yes |
Detectable Analytes | Salmonella |
Type | Salmonella Test Kit |
Product Code | Brand | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
13265639
|
Thermo Scientific™
R30858302 |
Wellcolex Colour Salmonella Kit | 200 Tests/Kit | 2°C to 8°C |
£1440.00
200 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11954042
|
Thermo Scientific™
R30858301 |
Wellcolex Colour Salmonella Kit | 50 Tests/Kit | 2°C to 8°C |
£550.00
50 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Rapidly detect and presumptively identify Salmonella from culture by latex agglutination using Thermo Scientific™ Wellcolex™ Color Salmonella Rapid Latex Agglutination Test Kit. Save up to two days' testing time over traditional screening procedures with this easy-to-use, cost-effective test. With its high positive and negative predictive values (PPV > 98.7%) and results in less than three minutes, laboratories can use this rapid screening test to eliminate Salmonella-negative samples from further testing, reducing the number of samples requiring confirmatory testing.
Use this rapid latex agglutination test for detection and presumptive serogrouping of Salmonella from Selenite Broth and solid culture media. Wellcolex Color Salmonella can also be used for the presumptive identification of Salmonella serogroups A, B, C, D, E, and G, and the Vi antigen using just two reagents.- Identifies major groups of Salmonella
- Utilizes advanced color technology for clear, easy-to-read species and/or serogroup identification
- Provides accurate, reliable results in less than three minutes
- Quickly eliminates negative samples when used as primary culture screening method
In the presence of homologous antigen one of the colors in the mixture will agglutinate, and the identity of the antigen is indicated by the color of the aggregated particles with a contrasting change in the color of the background. Each combination is easily distinguished from the others and from a negative result in which the particles remain in smooth grey‑brown suspension, and the occasional non‑specific result in which all the particles agglutinate into grey‑brown aggregates against a cleared background.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 8°C | |
Yes | |
Salmonella Test Kit | |
Salmonella |
Wellcolex Colour Salmonella Kit | |
Salmonella | |
Group A, B, C, D, E and G |