Learn More
Honeywell Safety Products™ Welding Cut Gloves
Heatproof and water-repellent cowhide are combined with cut and thermally resistant Kevlar™ for durability, resistance, and comfort. Honeywell Safety Products™ Welding Cut Gloves are designed with a protective cuff and an elasticized wrist for a good, protective fit.
Brand: Honeywell Safety Products™ 2049294-08
Code : NEW
Additional Details : Weight : 0.21600kg
Description
- Suitable for MIG and ARC welding in dry or light oily conditions with cut hazards.
- Water repellent, heatproof cowhide grain leather back.
- Water-repellent, heatproof velvet cowhide palm.
- Water repellent, heatproof split leather cuff to protect the forearm.
- U.S. assembly.
- Gunn cut, winged thumb with cord, Crispin type.
- Elasticated at the back of the hand.
- Kevlar™ cotton lining for comfort and thermal insulation. Outstanding comfort and flexibility.
Specifications
Cowgrain leather | |
Dexterity: 5 | |
Automotive, Building, Construction, Iron, Steel, Metal, Ship Building and Welding industries | |
Dupont™ Kevlar™ Aramid Fiber Cotton | |
Cowhide Leather | |
10Pair/PK | |
8 | |
Winged with yoke | |
39 cm | |
Palm and back: 0.8/1.1mm, Cuff: 1.1/1.3mm |
Split leather | |
Gunn | |
15 cm | |
Dupont™ Kevlar™ aramid fiber, Cotton | |
2 | |
Heat | |
2 | |
Gauntlet, Welding Type A, Crispin | |
CE CATEGORY II Intermediary risks, NF EN 420-03 EEC Basic standard, NF EN 388-03 Mechanical risks, NF EN 407-04 Thermical risks, EN 12477-02 Type A Welding risks, EC Category PPE 2, EC Certificate Number 0075/797/162/10/12/0763, EC Attestation Number 0075/797/162/10/12/0763 |