Learn More
Welch™ DirecTorr™ Gold Synthetic Pump Oil
Designed for excellent resistance when pumping corrosive gases or vapors
Brand: Welch™ U8995G-15
Code : A0
Additional Details : Weight : 0.01000kg
Description
Designed for excellent resistance when pumping corrosive gases or vapors
Enables excellent vacuum performance in both belt- and direct-driven vacuum pumps under harsh condition. Synthetic base stock has no aromatic compounds or sulfur. This purity inhibits varnishing, sludging and carbon buildup when pumping corrosives.
- Double-distilled synthetic hydrocarbon oil
- High stability in chemical environments with exceptionally low vapor pressure
- Miscible with conventional hydrocarbon oils and can be used without rebuilding the pump
- No additives or inhibitors
Specifications
Gold Vacuum Pump Oil | |
Recommended for ChemStar pump, belt-driven or direct-drive vacuum pumps | |
1 gal. |
Oil enables long service life and superior vacuum pump performance in applications where corrosive gases and vapors may be ingested | |
1 Ea. | |
3.78 L |