missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific Weighing Papers, White

The Weighing Paper is nitrogen-free for a very smooth surface and good sample recovery. It is strong enough to support dense material.

Brand:  Heathrow Scientific HS120118

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 16340706

  • £128.00 / Pack of 500
Estimated Shipment: 26-04-2024
to see stock.
Add to basket

Description

Description

  • Nitrogen-free
  • Disposable
Specifications

Specifications

6 in.
Weighing Papers
Paper
6 in.
15.2 cm
15.2 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers