missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Weighing Bottles

Manufactured from Borosilicate glass. DURAN™ Weighing Bottles come in several sizes.

Brand:  Duran™ 242112301

Code : A0

Additional Details : Weight : 0.42000kg

 View more versions of this product

Product Code. 11740964

  • £273.00 / Pack of 10


Estimated Shipment
05-06-2018
Add to basket

Description

Description

  • Low and tall forms
  • With grip stopper
  • Flat shape
  • Borosilicate glass
Specifications

Specifications

45mL
10Pack
70mm
High
Bottle, Weighing
Videos