missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ WDR5 (Human) Recombinant Protein
Human WDR5 partial ORF ( NP_060058, 16 a.a. - 120 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00011091-Q01.2ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: QPTPSSSATQSKPTPVKPNYALKFTLAGHTKAVSSVKFSPNGEWLASSSADKLIKIWGAYDGKFEKTISGHKLGISDVAWSSDSNLLVSASDDKTLKIWDVSSGK
Specifications
NP_060058 | |
WD repeat domain 5 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
WDR5 | |
GST |
11091 | |
Wheat germ expression system | |
2 μg | |
BIG-3, SWD3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |