Learn More
Solotec Scientific™ GP540 General Purpose Hood (previously WaySafe™ 3)
General purpose, quiet, portable laboratory hood/containment system. Solotec Scientific™ GP540 General Purpose Hood (previously WaySafe™ 3) draws air through a 300 series H14 HEPA filter before returning it to the lab.
Brand: Solotec Scientific™ GP540
Code : 04
Additional Details : Weight : 12.70000kg
Description
- Quiet, variable EC fan
- 300 series H14 HEPA filter
- Removable front panel
- Open base for easy positioning over fixed or heavy equipment
- Airflow is low enough to not disturb fine balances
- Vibration-free operation
- Flat packed and easily assembled