Learn More
Tocris Bioscience™ WAY 629 hydrochloride
Selective 5-HT2C agonist
Brand:  Tocris Bioscience™ 217310
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13460714
  • £106.65 / 10mg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
Selective 5-HT2C agonist. Ki values are 56, 2350, 1575 and 815 nM for human recombinant 5-HT2C, 5-HT2A, 5-HT6 and 5-HT7 receptors respectively. Stimulates intracellular Ca2+ mobilization in CHO cells expressing human 5-HT2C receptors (EC50 = 72 nM). Reduces food intake in rats.
Specifications
Specifications
WAY 629 hydrochloride
>98
10mg
WAY 629 hydrochloride
262.78