missing translation for 'onlineSavingsMsg'
Learn More

WAY 170523, Tocris Bioscience™

Potent and selective inhibitor of MMP-13

Brand:  RD Systems (Bio-Techne) 26331

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 1mg

 View more versions of this product

Product Code. 13441614

  • £116.13 / 1mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective inhibitor of MMP-13 (IC50 values are 17, 945, > 1000 and > 10000 nM for MMP-13, MMP-9, TACE and MMP-1 respectively).
Specifications

Chemical Identifiers

307002-73-9
613.685
n-2-4-benzyl 2-hydroxycarbamoyl-4,6-dimethylphenyl sulfamoyl phenoxy ethyl-1-benzofuran-2-carboxamide, d0ll8i, n-2-4-2-hydroxyamino carbonyl-4,6-dimethylphenyl phenylmethyl amino sulfonyl phenoxy ethyl-2-benzofurancarboxamide, way hplc, benzofuran-2-carboxylic acid 2-4-benzyl-2-hydroxycarbamoyl-4,6-dimethyl-phenyl-sulfamoyl-phenoxy-ethyl-amide, n-2-4-benzyl-2-hydroxycarbamoyl-4,6-dimethylphenyl sulfamoyl phenoxy ethyl-1-benzofuran-2-car
N-[2-[4-[benzyl-[2-(hydroxycarbamoyl)-4,6-dimethylphenyl]sulfamoyl]phenoxy]ethyl]-1-benzofuran-2-carboxamide
C33H31N3O7S
FARMEEAGJWMFSZ-UHFFFAOYSA-N
9830392
CC1=CC(=C(C(=C1)C(=O)NO)N(CC2=CC=CC=C2)S(=O)(=O)C3=CC=C(C=C3)OCCNC(=O)C4=CC5=CC=CC=C5O4)C

Specifications

1mg
307002-73-9
n-2-4-benzyl 2-hydroxycarbamoyl-4,6-dimethylphenyl sulfamoyl phenoxy ethyl-1-benzofuran-2-carboxamide, d0ll8i, n-2-4-2-hydroxyamino carbonyl-4,6-dimethylphenyl phenylmethyl amino sulfonyl phenoxy ethyl-2-benzofurancarboxamide, way hplc, benzofuran-2-carboxylic acid 2-4-benzyl-2-hydroxycarbamoyl-4,6-dimethyl-phenyl-sulfamoyl-phenoxy-ethyl-amide, n-2-4-benzyl-2-hydroxycarbamoyl-4,6-dimethylphenyl sulfamoyl phenoxy ethyl-1-benzofuran-2-car
CC1=CC(=C(C(=C1)C(=O)NO)N(CC2=CC=CC=C2)S(=O)(=O)C3=CC=C(C=C3)OCCNC(=O)C4=CC5=CC=CC=C5O4)C
613.685
613.68
WAY 170523
C33H31N3O7S
FARMEEAGJWMFSZ-UHFFFAOYSA-N
N-[2-[4-[benzyl-[2-(hydroxycarbamoyl)-4,6-dimethylphenyl]sulfamoyl]phenoxy]ethyl]-1-benzofuran-2-carboxamide
9830392
>99%
Videos
Documents